1-(4-Methoxybenzyl)pyrrolidin-2-one 95+%

Lieferant: Apollo Scientific

OR346540-500MG
APOSOR346540-500MGEA 420 EUR
APOSOR346540-500MG
1-(4-Methoxybenzyl)pyrrolidin-2-one 95+%
1-(4-Methoxybenzyl)pyrrolidin-2-one
Formel: C₁₂H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01960843
CAS-Nummer: 60737-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR