4-Isopropylpicolinic acid 95+%
Lieferant: Apollo Scientific
OR346542-1G
APOSOR346542-1GEA
751
EUR
APOSOR346542-1G
4-Isopropylpicolinic acid 95+%
4-Isopropylpicolinic acid
Formel:
C₉H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437963 CAS-Nummer: 83282-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...