04-Amino-2-ethoxybenzonitrile 95+%
Lieferant: Apollo Scientific
OR346544-1G
APOSOR346544-1GEA
497
EUR
APOSOR346544-1G
04-Amino-2-ethoxybenzonitrile 95+%
04-Amino-2-ethoxybenzonitrile
Formel:
C₉H₁₀N₂O Molecular Weight: 162,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20694465 CAS-Nummer: 609783-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...