1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%

Lieferant: Apollo Scientific

OR346547-1G
APOSOR346547-1GEA 253 EUR
APOSOR346547-1G
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone
Formel: C₁₄H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12404914
CAS-Nummer: 1215576-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR