1-[6-(3-Methoxyphenyl)pyridin-3-yl]ethanone 95+%

Lieferant: Apollo Scientific

OR346549-1G
APOSOR346549-1GEA 265 EUR
APOSOR346549-1G
1-[6-(3-Methoxyphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(3-Methoxyphenyl)pyridin-3-yl]ethanone
Formel: C₁₄H₁₃NO₂
Molecular Weight: 227.27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404915
CAS-Nummer: 1215576-39-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR