1-[6-(4-Methylphenyl)pyridin-3-yl]ethanone 95+%

Lieferant: Apollo Scientific

Synonyme: 1-(6-p-Tolylpyridin-3-yl)ethanone

OR346550-1G
APOSOR346550-1GEA 265 EUR
APOSOR346550-1G
1-[6-(4-Methylphenyl)pyridin-3-yl]ethanone 95+%
1-[6-(4-Methylphenyl)pyridin-3-yl]ethanone
Formel: C₁₄H₁₃NO
Molecular Weight: 211,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404918
CAS-Nummer: 1216790-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR