3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine 95+%

Lieferant: Apollo Scientific

OR346555-1G
APOSOR346555-1GEA 0 EUR
APOSOR346555-1G
3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine 95+%
3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine
Formel: C₉H₁₀BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29918682
CAS-Nummer: 936844-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR