4-Bromo-2-methylbenzamide 95+%

Lieferant: Apollo Scientific

OR346556-1G
APOSOR346556-1GEA 171 EUR
APOSOR346556-1G
4-Bromo-2-methylbenzamide 95+%
4-Bromo-2-methylbenzamide
Formel: C₈H₈BrNO
Molecular Weight: 214.059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12096764
CAS-Nummer: 868776-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR