4-(Methylsulphonyl)benzoyl chloride 95+%

Lieferant: Apollo Scientific

OR346557-1G
APOSOR346557-1GEA 0 EUR
APOSOR346557-1G
4-(Methylsulphonyl)benzoyl chloride 95+%
4-(Methylsulphonyl)benzoyl chloride
Formel: C₈H₇ClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD11169646
CAS-Nummer: 40913-92-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR