4-(tert-Butyl)pyridine 1-oxide 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(tert-Butyl)pyridine N-oxide, 4-(2-Methyl-2-propanyl)pyridine 1-oxide, 4-tert-Butyl-1-oxypyridine, 4-tert-Butylpyridine 1-oxide

OR346558-1G
APOSOR346558-1GEA 136 EUR
APOSOR346558-1G
4-(tert-Butyl)pyridine 1-oxide 95+%
4-tert-Butylpyridine-1-oxide
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082431
CAS-Nummer: 23569-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR