1-(2-Methoxyphenyl)propan-1-one 95+%

Lieferant: Apollo Scientific

OR346562-1G
APOSOR346562-1GEA 294 EUR
APOSOR346562-1G
1-(2-Methoxyphenyl)propan-1-one 95+%
1-(2-Methoxyphenyl)propan-1-one
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048613
CAS-Nummer: 5561-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR