2-Bromo-1-(3-methoxyphenyl)propan-1-one 95+%

Lieferant: Apollo Scientific

OR346564-1G
APOSOR346564-1GEA 598 EUR
APOSOR346564-1G
2-Bromo-1-(3-methoxyphenyl)propan-1-one 95+%
2-Bromo-1-(3-methoxyphenyl)propan-1-one
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031564
CAS-Nummer: 21726-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR