2-Bromo-1-(4-methoxyphenyl)propan-1-one 95+%

Lieferant: Apollo Scientific

OR346565-1G
APOSOR346565-1GEA 306 EUR
APOSOR346565-1G
2-Bromo-1-(4-methoxyphenyl)propan-1-one 95+%
2-Bromo-1-(4-methoxyphenyl)propan-1-one
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243,097 g/mol
Schmelzpunkt: 66…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00790769
CAS-Nummer: 21086-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR