4-Chloro-3-methoxybenzenesulphonyl chloride 95+%

Lieferant: Apollo Scientific

OR346566-1G
APOSOR346566-1GEA 751 EUR
APOSOR346566-1G
4-Chloro-3-methoxybenzenesulphonyl chloride 95+%
4-Chloro-3-methoxybenzenesulphonyl chloride
Formel: C₇H₆Cl₂O₃S
Molecular Weight: 241,092 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12402639
CAS-Nummer: 942199-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR