5-Amino-2-chlorobenzoic acid isopropyl ester 95+%

Lieferant: Apollo Scientific

OR346571-1G
APOSOR346571-1GEA 102 EUR
APOSOR346571-1G
5-Amino-2-chlorobenzoic acid isopropyl ester 95+%
5-Amino-2-chlorobenzoic acid isopropyl ester
Formel: C₁₀H₁₂ClNO₂
Molecular Weight: 213,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05133182
CAS-Nummer: 86819-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR