7-Methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one 95+%

Lieferant: Apollo Scientific

OR346572-1G
APOSOR346572-1GEA 369 EUR
APOSOR346572-1G
7-Methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one 95+%
7-Methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
Formel: C₇H₆N₂OS
Molecular Weight: 166,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00552712
CAS-Nummer: 700-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR