4,5-Dimethyl-2-phenyl-1,3-thiazole 95+%

Lieferant: Apollo Scientific

OR346573-1G
APOSOR346573-1GEA 0 EUR
APOSOR346573-1G
4,5-Dimethyl-2-phenyl-1,3-thiazole 95+%
4,5-Dimethyl-2-phenyl-1,3-thiazole
Formel: C₁₁H₁₁NS
Molecular Weight: 189,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272329
CAS-Nummer: 1826-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR