3,4-Dihydro-2H-isoquinoline-1-thione 95+%

Lieferant: Apollo Scientific

OR346575-1G
APOSOR346575-1GEA 0 EUR
APOSOR346575-1G
3,4-Dihydro-2H-isoquinoline-1-thione 95+%
3,4-Dihydro-2H-isoquinoline-1-thione
Formel: C₉H₉NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00126731
CAS-Nummer: 6552-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR