4-Thiazol-2-ylbenzaldehyde 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(Thiazol-2-yl)benzaldehyde

OR346576-1G
APOSOR346576-1GEA 43.5 EUR
APOSOR346576-1G
4-Thiazol-2-ylbenzaldehyde 95+%
4-Thiazol-2-ylbenzaldehyde
Formel: C₁₀H₇NOS
Molecular Weight: 189,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797780
CAS-Nummer: 198904-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR