4,6-Dichloro-pyrimidine-5-carbonyl chloride 95+%

Lieferant: Apollo Scientific

OR346585-1G
APOSOR346585-1GEA 700 EUR
APOSOR346585-1G
4,6-Dichloro-pyrimidine-5-carbonyl chloride 95+%
4,6-Dichloro-pyrimidine-5-carbonyl chloride
Formel: C₅HCl₃N₂O
Molecular Weight: 211,433 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698804
CAS-Nummer: 87600-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR