5-Methylpyridine-3-sulphonyl chloride 95+%

Lieferant: Apollo Scientific

OR346586-1G
APOSOR346586-1GEA 497 EUR
APOSOR346586-1G
5-Methylpyridine-3-sulphonyl chloride 95+%
5-Methylpyridine-3-sulphonyl chloride
Formel: C₆H₆ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD13188992
CAS-Nummer: 166337-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR