Ethyl-4-amino-3-chlorbenzoat 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-amino-3-chlorobenzoate, 4-Amino-3-chlorobenzoic acid ethyl ester

OR346587-5G
APOSOR346587-5GEA 58 EUR
APOSOR346587-5G
Ethyl-4-amino-3-chlorbenzoat 95+%
Ethyl-4-amino-3-chlorbenzoat
Formel: C₉H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06411007
CAS-Nummer: 82765-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR