1-(4-Aminophenyl)-cyclobutanecarboxylic acid methyl ester 95+%
Lieferant: Apollo Scientific
OR346589-1G
APOSOR346589-1GEA
751
EUR
APOSOR346589-1G
1-(4-Aminophenyl)-cyclobutanecarboxylic acid methyl ester 95+%
1-(4-Aminophenyl)-cyclobutanecarboxylic acid methyl ester
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,253 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24627542 CAS-Nummer: 1215017-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...