1-Pyridin-3-yl-cyclopentanecarbonitrile 95+%
Lieferant: Apollo Scientific
OR346599-1G
APOSOR346599-1GEA
623
EUR
APOSOR346599-1G
1-Pyridin-3-yl-cyclopentanecarbonitrile 95+%
1-Pyridin-3-yl-cyclopentanecarbonitrile
Formel:
C₁₁H₁₂N₂ Molecular Weight: 172,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15144542 CAS-Nummer: 143619-68-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...