4-(2-Chloro-ethyl)benzamide 95+%

Lieferant: Apollo Scientific

OR346600-1G
APOSOR346600-1GEA 623 EUR
APOSOR346600-1G
4-(2-Chloro-ethyl)benzamide 95+%
4-(2-Chloro-ethyl)benzamide
Formel: C₉H₁₀ClNO
Molecular Weight: 183.635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20637141
CAS-Nummer: 181115-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR