5-Chloro-3-isopropyl-1-methyl-1H-pyrazole 95+%

Lieferant: Apollo Scientific

OR346603-1G
APOSOR346603-1GEA 623 EUR
APOSOR346603-1G
5-Chloro-3-isopropyl-1-methyl-1H-pyrazole 95+%
5-Chloro-3-isopropyl-1-methyl-1H-pyrazole
Formel: C₇H₁₁ClN₂
Molecular Weight: 158,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05860084
CAS-Nummer: 29938-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR