3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95+%
Lieferant: Apollo Scientific
OR346605-1G
APOSOR346605-1GEA
84.1
EUR
APOSOR346605-1G
3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95+%
3-Bromo[1,2,4]triazolo[4,3-a]pyridine
Formel:
C6H4BrN3 Molecular Weight: 198,0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111699 CAS-Nummer: 4922-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...