[2-(1-Pyrrolidinyl)-3-pyridinyl]methanol 95+%
Lieferant: Apollo Scientific
OR346607-1G
APOSOR346607-1GEA
340
EUR
APOSOR346607-1G
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanol 95+%
[2-(1-Pyrrolidinyl)-3-pyridinyl]methanol
Formel:
C₁₀H₁₄N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664425 CAS-Nummer: 690632-85-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...