4-Phenyl-1H-pyrrole-3-carboxylic acid 95+%

Lieferant: Apollo Scientific

OR346623-1G
APOSOR346623-1GEA 192 EUR
APOSOR346623-1G
4-Phenyl-1H-pyrrole-3-carboxylic acid 95+%
4-Phenyl-1H-pyrrole-3-carboxylic acid
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06411253
CAS-Nummer: 132040-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR