1-Iodo-2-(methylsulphinyl)benzene 95+%

Lieferant: Apollo Scientific

OR346624-1G
APOSOR346624-1GEA 623 EUR
APOSOR346624-1G
1-Iodo-2-(methylsulphinyl)benzene 95+%
1-Iodo-2-(methylsulphinyl)benzene
Formel: C₇H₇IOS
Molecular Weight: 266,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18089236
CAS-Nummer: 71545-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR