(Benzofuran-2-yl)acetonitrile 95+%

Lieferant: Apollo Scientific

OR346625-1G OR346625-500MG
APOSOR346625-1GEA 876 EUR
APOSOR346625-1G APOSOR346625-500MG
(Benzofuran-2-yl)acetonitrile 95+%
(Benzofuran-2-yl)acetonitrile
Formel: C₁₀H₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00052739
CAS-Nummer: 95-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR