2-Methylfuran-3-carboxamide 95+%

Lieferant: Apollo Scientific

OR346629-1G
APOSOR346629-1GEA 318 EUR
APOSOR346629-1G
2-Methylfuran-3-carboxamide 95+%
2-Methylfuran-3-carboxamide
Formel: C₆H₇NO₂
Molecular Weight: 125.125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11643537
CAS-Nummer: 22727-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR