3-Methylfuran-2-carboxamide 95+%

Lieferant: Apollo Scientific

OR346630-1G
APOSOR346630-1GEA 369 EUR
APOSOR346630-1G
3-Methylfuran-2-carboxamide 95+%
3-Methylfuran-2-carboxamide
Formel: C₆H₇NO₂
Molecular Weight: 125.125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11643939
CAS-Nummer: 84374-70-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR