3-Benzyloxyisothiazole 95+%

Lieferant: Apollo Scientific

OR346631-1G
APOSOR346631-1GEA 497 EUR
APOSOR346631-1G
3-Benzyloxyisothiazole 95+%
3-Benzyloxyisothiazole
Formel: C₁₀H₉NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD27926230
CAS-Nummer: 60666-83-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR