5-Brom-2,3-dihydro-1-benzofuran-7-carbonsäure 95+%

Lieferant: Apollo Scientific

Synonyme: 5-Bromocoumaran-7-carboxylic Acid

OR346634-1G
APOSOR346634-1GEA 184 EUR
APOSOR346634-1G
5-Brom-2,3-dihydro-1-benzofuran-7-carbonsäure 95+%
5-Brom-2,3-dihydro-1-benzofuran-7-carbonsäure
Formel: C₉H₇BrO₃
Molecular Weight: 243,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191391
CAS-Nummer: 41177-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR