3-Ethylacetylaceton 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Ethyl-2,4-pentanedione, 3-Ethyl-4-hydroxy-3-penten-3-one

OR346636-10G
APOSOR346636-10GEA 197 EUR
APOSOR346636-10G
3-Ethylacetylaceton 95+%
3-Ethylacetylaceton
Formel: C₇H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00008764
CAS-Nummer: 1540-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR