6-(2-Methylphenyl)nicotinic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 6-(o-Tolyl)nicotinic acid, 6-o-Tolylnicotinic acid

OR346644-1G
APOSOR346644-1GEA 318 EUR
APOSOR346644-1G
6-(2-Methylphenyl)nicotinic acid 95+%
6-(2-Methylphenyl)nicotinic acid
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410397
CAS-Nummer: 175153-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR