6-Hydroxymethylpyridine-2-carbonitrile 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Hydroxymethyl)picolinonitrile

OR346651-1G
APOSOR346651-1GEA 200 EUR
APOSOR346651-1G
6-Hydroxymethylpyridine-2-carbonitrile 95+%
6-Hydroxymethylpyridine-2-carbonitrile
Formel: C₇H₆N₂O
Molecular Weight: 134,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367926
CAS-Nummer: 50501-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR