3-(1-Hydroxy-1-methylethyl)benzoic acid 95+%

Lieferant: Apollo Scientific

OR346657-1G
APOSOR346657-1GEA 751 EUR
APOSOR346657-1G
3-(1-Hydroxy-1-methylethyl)benzoic acid 95+%
3-(1-Hydroxy-1-methylethyl)benzoic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260812
CAS-Nummer: 40912-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR