(5-Bromobenzofuran-2-yl)acetic acid 95+%

Lieferant: Apollo Scientific

OR346660-1G
APOSOR346660-1GEA 760 EUR
APOSOR346660-1G
(5-Bromobenzofuran-2-yl)acetic acid 95+%
(5-Bromobenzofuran-2-yl)acetic acid
Formel: C₁₀H₇BrO₃
Molecular Weight: 255,065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11574519
CAS-Nummer: 84102-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR