(2-Iodophenyl)acetyl choride 95+%

Lieferant: Apollo Scientific

OR346673-1G
APOSOR346673-1GEA 366 EUR
APOSOR346673-1G
(2-Iodophenyl)acetyl choride 95+%
(2-Iodophenyl)acetyl choride
Formel: C₈H₆ClIO
Molecular Weight: 280,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025057
CAS-Nummer: 62300-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR