2-(Bromomethyl)benzaldehyde 95+%

Lieferant: Apollo Scientific

OR346678-1G
APOSOR346678-1GEA 0 EUR
APOSOR346678-1G
2-(Bromomethyl)benzaldehyde 95+%
2-(Bromomethyl)benzaldehyde
Formel: C₈H₇BrO
Lagertemperatur: Kühlschrank
MDL: MFCD17014845
CAS-Nummer: 60633-91-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR