2-(2-Iodophenyl)acetamide 95+%

Lieferant: Apollo Scientific

OR346701-1G
APOSOR346701-1GEA 751 EUR
APOSOR346701-1G
2-(2-Iodophenyl)acetamide 95+%
2-(2-Iodophenyl)acetamide
Formel: C₈H₈INO
Molecular Weight: 261,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00099253
CAS-Nummer: 195456-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR