5-(Chloromethyl)-3-isopropylisoxazole 95+%

Lieferant: Apollo Scientific

OR346708-1G
APOSOR346708-1GEA 318 EUR
APOSOR346708-1G
5-(Chloromethyl)-3-isopropylisoxazole 95+%
5-(Chloromethyl)-3-isopropylisoxazole
Formel: C₇H₁₀ClNO
Molecular Weight: 159.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09701716
CAS-Nummer: 64988-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR