1-(3-Phenylisoxazol-5-yl)-ethanol 90+%

Lieferant: Apollo Scientific

OR346711-1G
APOSOR346711-1GEA 751 EUR
APOSOR346711-1G
1-(3-Phenylisoxazol-5-yl)-ethanol 90+%
1-(3-Phenylisoxazol-5-yl)-ethanol
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08443700
CAS-Nummer: 14776-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR