4-(Pyrrol-1-yl)benzaldehyde 95+%

Lieferant: Apollo Scientific

OR346717-5G
APOSOR346717-5GEA 111 EUR
APOSOR346717-5G
4-(Pyrrol-1-yl)benzaldehyde 95+%
4-(Pyrrol-1-yl)benzaldehyde
Formel: C₁₁H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03822126
CAS-Nummer: 23351-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR