6-Iodonicotinic acid 95+%

Lieferant: Apollo Scientific

OR346718-1G
APOSOR346718-1GEA 369 EUR
APOSOR346718-1G
6-Iodonicotinic acid 95+%
6-Iodonicotinic acid
Formel: C₆H₄INO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03426924
CAS-Nummer: 13054-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR