2-Bromo-4-chloro-5-methoxybenzoic acid 95+%

Lieferant: Apollo Scientific

OR346720-1G
APOSOR346720-1GEA 369 EUR
APOSOR346720-1G
2-Bromo-4-chloro-5-methoxybenzoic acid 95+%
2-Bromo-4-chloro-5-methoxybenzoic acid
Formel: C₈H₆BrClO₃
Molecular Weight: 265,488 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28967300
CAS-Nummer: 1435479-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR