2,2-Dichloro-2-phenylacetonitrile 97%

Lieferant: Apollo Scientific

OR3472-1G OR3472-5G
APOSOR3472-1GEA 0 EUR
APOSOR3472-1G APOSOR3472-5G
2,2-Dichloro-2-phenylacetonitrile 97%
2,2-Dichloro-2-phenylacetonitrile
Formel: C₈H₅Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09702429
CAS-Nummer: 40626-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR