6-Chlor-5-nitronicotinsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR3496-1G
APOSOR3496-1GEA
21.9
EUR
APOSOR3496-1G
6-Chlor-5-nitronicotinsäure 95%
6-Chlor-5-nitronicotinsäure
Formel:
C₆H₃ClN₂O₄ Molecular Weight: 202,5520 g/mol Schmelzpunkt: 99…102 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09027297 CAS-Nummer: 7477-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...